Anti-SLC6A8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-12124R-A350
BOSSBS-12124R-A350EA 10450 NOK
BOSSBS-12124R-A350
Anti-SLC6A8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
The GAT1 gene family includes sodium- and chloride-dependent plasma membrane transporters for neurotransmitters, metabolites and osmolites, which couple substrate flux to transmembrane electrochemical gradients. PROT (Sodium-dependent proline transporter), also known as Solute carrier family 6 member 7, is a 636 amino acid multi-pass membrane protein that is a GAT1 family member specifically expressed in regions of the brain. PROT terminates the action of proline by its high affinity sodium/chloride-dependent reuptake into pre-synaptic terminals. Enriched in glutamatergic synaptic terminals,More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol SLC6A8
Antigen-navn Solute Carrier Family 6 (neurotransmitter transporter creatine) Member 8
Antigen synonymer Solute Carrier Family 6|CRT|solute carrier family 6 member 8|MGC87396|creatine transporter SLC6A8|CT1|creatine transporter 1|sodium- and chloride-dependent creatine transporter 1|neurotransmitter transporter creatine member 8|CRTR
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Flowcytometri Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR