Anti-SLC6A11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Leverandør: Bioss
BS-12124R-A647
BOSSBS-12124R-A647EA 10450 NOK
BOSSBS-12124R-A647
Anti-SLC6A11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
The GAT1 gene family includes sodium- and chloride-dependent plasma membrane transporters for neurotransmitters, metabolites and osmolites, which couple substrate flux to transmembrane electrochemical gradients. PROT (Sodium-dependent proline transporter), also known as Solute carrier family 6 member 7, is a 636 amino acid multi-pass membrane protein that is a GAT1 family member specifically expressed in regions of the brain. PROT terminates the action of proline by its high affinity sodium/chloride-dependent reuptake into pre-synaptic terminals. Enriched in glutamatergic synaptic terminals,More Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol SLC6A11
Antigen-navn Solute Carrier Family 6 (Neurotransmitter Transporter), Member 11
Antigen synonymer Sodium-dependent proline transporter|Slc6a7|Solute carrier family 6 member 7.|SC6A7_HUMAN
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 647
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Flowcytometri Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C