Anti-SLC6A7/PROT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-12124R-A680
BOSSBS-12124R-A680EA 10425 NOK
BOSSBS-12124R-A680
Anti-SLC6A7/PROT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
The GAT1 gene family includes sodium- and chloride-dependent plasma membrane transporters for neurotransmitters, metabolites and osmolites, which couple substrate flux to transmembrane electrochemical gradients. PROT (Sodium-dependent proline transporter), also known as Solute carrier family 6 member 7, is a 636 amino acid multi-pass membrane protein that is a GAT1 family member specifically expressed in regions of the brain. PROT terminates the action of proline by its high affinity sodium/chloride-dependent reuptake into pre-synaptic terminals. Enriched in glutamatergic synaptic terminals,More Product Information
Bestill nå
SPESIFIKASJONER
Antigen-navn SLC6A7/PROT
Antigen synonymer Solute carrier family 6 member 7|Sodium-dependent proline transporter|Slc6a7|SC6A7_HUMAN
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 151-260/636
Immunogen KLH conjugated synthetic peptide derived from human SLC6A7/PROT
Skjema Liquid
Molekylvekt 71 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC