Anti-SLC6A16 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-12124R-FITC
BOSSBS-12124R-FITCEA 10550 NOK
BOSSBS-12124R-FITC
Anti-SLC6A16 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
The GAT1 gene family includes sodium- and chloride-dependent plasma membrane transporters for neurotransmitters, metabolites and osmolites, which couple substrate flux to transmembrane electrochemical gradients. PROT (Sodium-dependent proline transporter), also known as Solute carrier family 6 member 7, is a 636 amino acid multi-pass membrane protein that is a GAT1 family member specifically expressed in regions of the brain. PROT terminates the action of proline by its high affinity sodium/chloride-dependent reuptake into pre-synaptic terminals. Enriched in glutamatergic synaptic terminals,More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol SLC6A16
Antigen-navn Solute Carrier Family 6 (Neurotransmitter Transporter), Member 16
Antigen synonymer Sodium-dependent proline transporter|Slc6a7|Solute carrier family 6 member 7.|SC6A7_HUMAN
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Flowcytometri Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR