Anti-PPP1R9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-12146R-A350
BOSSBS-12146R-A350EA 10450 NOK
BOSSBS-12146R-A350
Anti-PPP1R9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Seems to act as a scaffold protein in multiple signaling pathways. Modulates excitatory synaptic transmission and dendritic spine morphology. Binds to actin filaments (F-actin) and shows cross-linking activity. Binds along the sides of the F-actin. May play an important role in linking the actin cytoskeleton to the plasma membrane at the synaptic junction. Believed to target protein phosphatase 1/PP1 to dendritic spines, which are rich in F-actin, and regulates its specificity toward ion channels and other substrates, such as AMPA-type and NMDA-type glutamate receptors. Plays a role inMore Product Information