Anti-PPP1R9 Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-12146R-CY5
BOSSBS-12146R-CY5EA 10550 NOK
BOSSBS-12146R-CY5
Anti-PPP1R9 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Seems to act as a scaffold protein in multiple signaling pathways. Modulates excitatory synaptic transmission and dendritic spine morphology. Binds to actin filaments (F-actin) and shows cross-linking activity. Binds along the sides of the F-actin. May play an important role in linking the actin cytoskeleton to the plasma membrane at the synaptic junction. Believed to target protein phosphatase 1/PP1 to dendritic spines, which are rich in F-actin, and regulates its specificity toward ion channels and other substrates, such as AMPA-type and NMDA-type glutamate receptors. Plays a role inMore Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol PPP1R9
Antigen-navn Protein Phosphatase 1, Regulatory Subunit 9B
Environmentally Preferable
Antigen synonymer PPP1R9B |Neurabin 2|SPN|PPP1R6|PPP1R9|Spinophilin
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 84687
Isotype IgG
ImmunoKjemi Yes
Immunogen 360-410/815
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C