Anti-PSMA4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-12260R-A488
BOSSBS-12260R-A488EA 10450 NOK
BOSSBS-12260R-A488
Anti-PSMA4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
The proteasome represents a large protein complex that exists inside all eukaryotes and archaea, and in some bacteria. The main function of proteasomes is to degrade unnecessary or damaged proteins by proteolysis. The most common form of the proteasome, known as the 26S Proteasome, contains one 20S Proteasome core particle structure and two 19S regulatory caps. The 20S Proteasome core is hollow and forms an enclosed cavity, where proteins are degraded, as well as openings at the two ends to allow the target protein to enter. The 20S Proteasome core particle contains many subunits, dependingMore Product Information