Anti-CEPT1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-12284R
BOSSBS-12284REA 8500 NOK
BOSSBS-12284R
Anti-CEPT1 Rabbit Polyclonal Antibody
Antistoffer
Cholinephosphotransferase catalyses the final step in the synthesis of phosphatidylcholine by the transfer of phosphocholine from CDP-choline to diacylglycerol. The synthesis of phosphatidylethanolamine by ethanolaminephosphotransferase occurs using an analogous reaction. This gene codes for a choline/ethanolaminephosphotransferase. The protein can synthesize either choline- or ethanolamine- containing phospholipids. Two alternatively spliced transcripts encoding the same isoform have been identified. [provided by RefSeq, Jul 2008].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Bestill nå

SPESIFIKASJONER

Antigensymbol CEPT1
Antigen-navn Choline/ethanolamine phosphotransferase 1
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 10390
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR