Anti-CEPT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-12284R-A750
BOSSBS-12284R-A750EA 10425 NOK
BOSSBS-12284R-A750
Anti-CEPT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Cholinephosphotransferase catalyses the final step in the synthesis of phosphatidylcholine by the transfer of phosphocholine from CDP-choline to diacylglycerol. The synthesis of phosphatidylethanolamine by ethanolaminephosphotransferase occurs using an analogous reaction. This gene codes for a choline/ethanolaminephosphotransferase. The protein can synthesize either choline- or ethanolamine- containing phospholipids. Two alternatively spliced transcripts encoding the same isoform have been identified.

Type: Primary
Antigen: CEPT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR