Anti-CEPT1 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-12284R-CY7
BOSSBS-12284R-CY7EA 10550 NOK
BOSSBS-12284R-CY7
Anti-CEPT1 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Cholinephosphotransferase catalyses the final step in the synthesis of phosphatidylcholine by the transfer of phosphocholine from CDP-choline to diacylglycerol. The synthesis of phosphatidylethanolamine by ethanolaminephosphotransferase occurs using an analogous reaction. This gene codes for a choline/ethanolaminephosphotransferase. The protein can synthesize either choline- or ethanolamine- containing phospholipids. Two alternatively spliced transcripts encoding the same isoform have been identified. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200