Anti-GSC Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-12352R-A488
BOSSBS-12352R-A488EA 10450 NOK
BOSSBS-12352R-A488
Anti-GSC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Goosecoid is a homeodomain transcription factor with DNA binding specificity identical to that of the anterior morphogen 'bicoid' in Drosophila.During mouse embryogenesis, GSC influences development of the lower mandible and its associated musculature, including the tongue, the nasal cavity, and the nasal pits, as well as components of the inner ear and the external auditory meatus. The GSC gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSC
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR