Anti-GSC Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-12352R-CY3
BOSSBS-12352R-CY3EA 10550 NOK
BOSSBS-12352R-CY3
Anti-GSC Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Goosecoid is a homeodomain transcription factor with DNA binding specificity identical to that of the anterior morphogen 'bicoid' in Drosophila.During mouse embryogenesis, GSC influences development of the lower mandible and its associated musculature, including the tongue, the nasal cavity, and the nasal pits, as well as components of the inner ear and the external auditory meatus. The GSC gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSC
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GSC
Antigen-navn Goosecoid Homeobox
Environmentally Preferable
Antigen synonymer SAMS
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy3®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 145258
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR