Anti-HO-2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-1238R-A680
BOSSBS-1238R-A680EA 10425 NOK
BOSSBS-1238R-A680
Anti-HO-2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Heme oxygenase 2 could be implicated in the production of carbon monoxide in brain where it could act as a neurotransmitter.

Type: Primary
Antigen: HO-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-150/316
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR