Anti-HMOX2 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-1238R-CY3
BOSSBS-1238R-CY3EA 10550 NOK
BOSSBS-1238R-CY3
Anti-HMOX2 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Heme oxygenase 2 could be implicated in the production of carbon monoxide in brain where it could act as a neurotransmitter.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMOX2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR