Anti-HMOX2 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-1238R-CY5.5
BOSSBS-1238R-CY5.5EA 10550 NOK
BOSSBS-1238R-CY5.5
Anti-HMOX2 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Heme oxygenase 2 could be implicated in the production of carbon monoxide in brain where it could act as a neurotransmitter.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMOX2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HMOX2
Environmentally Preferable
Antigen-navn Heme Oxidase 2
Antigen synonymer HO-2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 3163
Isotype IgG
ImmunoKjemi Yes
Immunogen 40-90/316
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR