Anti-MAML2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-12398R-A647
BOSSBS-12398R-A647EA 10450 NOK
BOSSBS-12398R-A647
Anti-MAML2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
The three MAML genes are widely expressed in adult tissues but exhibit distinct expression patterns in mouse early spinal cord development. All MAML proteins localize to nuclear bodies, share a conserved basic domain in their N termini that binds to the ankyrin repeat domain of Notch, and contain a transcriptional activation domain in their C termini. MAML3 acts as a transcriptional coactivator for NOTCH proteins and has been shown to amplify NOTCH-induced transcription of HES1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAML2
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol MAML2
Environmentally Preferable
Antigen-navn Mastermind-Like 2
Antigen synonymer Mastermind-Like 3 Protein|MAM3|MAM-3|MAM2|MLL-MAML2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 647
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR