Anti-MAML2 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-12398R-CY7
BOSSBS-12398R-CY7EA 10550 NOK
BOSSBS-12398R-CY7
Anti-MAML2 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
The three MAML genes are widely expressed in adult tissues but exhibit distinct expression patterns in mouse early spinal cord development. All MAML proteins localize to nuclear bodies, share a conserved basic domain in their N termini that binds to the ankyrin repeat domain of Notch, and contain a transcriptional activation domain in their C termini. MAML3 acts as a transcriptional coactivator for NOTCH proteins and has been shown to amplify NOTCH-induced transcription of HES1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAML2
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR