Anti-MAML2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-12398R-FITC
BOSSBS-12398R-FITCEA 10550 NOK
BOSSBS-12398R-FITC
Anti-MAML2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
The three MAML genes are widely expressed in adult tissues but exhibit distinct expression patterns in mouse early spinal cord development. All MAML proteins localize to nuclear bodies, share a conserved basic domain in their N termini that binds to the ankyrin repeat domain of Notch, and contain a transcriptional activation domain in their C termini. MAML3 acts as a transcriptional coactivator for NOTCH proteins and has been shown to amplify NOTCH-induced transcription of HES1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAML2
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol MAML2
Environmentally Preferable
Antigen-navn Mastermind-Like 2
Antigen synonymer Mastermind-Like 3 Protein|MAM3|MAM-3|MAM2|MLL-MAML2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR