Anti-SNX4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-12406R-A680
BOSSBS-12406R-A680EA 10425 NOK
BOSSBS-12406R-A680
Anti-SNX4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinctMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn SNX4
Antigen synonymer SNX4_HUMAN|SNX 4|Sorting nexin 4|snx4|ATG24B|Sorting nexin-4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Vert Rabbit
Gen-ID 8723
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 351-450/450
Immunogen KLH conjugated synthetic peptide derived from human SNX4
Skjema Liquid
Molekylvekt 52 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR