Anti-DOCK5 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-12433R
BOSSBS-12433REA 12775 NOK
BOSSBS-12433R
Anti-DOCK5 Rabbit Polyclonal Antibody
Antistoffer
DOCK 5 is a 1,870 amino acid protein belonging to the DOCK family of cytokinesis-regulating proteins. This cytoplasmic peripheral membrane protein activates Rac 1 and Rac 2 small GTPases, while presumably acting as a guanine nucleotide exchange factor (GEF), which exchanges bound GDP for free GTP. DOCK 5 contains one DHR-1 (CZH-1) domain, one DHR-2 (CZH-2) domain and one SH3 domain. The DHR-2 domain is a putative GEF activity mediator. In mice, spontaneous mutation of the gene encoding DOCK 5 leads to deletion of the DHR-1 domain, which functions to bind phospholipids and assists in protein-More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol DOCK5
Antigen-navn dedicator of cytokinesis 5
Environmentally Preferable
Antigen synonymer DKFZp781J211|DKFZp779M164|DKFZp451J181|DOCK5_HUMAN.|Dedicator of cytokinesis protein 5|Dedicator of cytokinesis 5|Dock 5
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 80005
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR