Anti-APC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Bioss
BS-12485R-HRP
BOSSBS-12485R-HRPEA 10550 NOK
BOSSBS-12485R-HRP
Anti-APC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The adenomatous polyposis syndromes, familial adenomatous polyposis (FAP) and Gardner’s syndrome (GS), are characterized by numerous adenomatous polyps throughout the entire colon. These polyps invariably progress to colon cancer in addition to other extracolonic manifestations. The cloning of the APC gene revealed a ubiquitously expressed protein, 2843 amino acids in length, which is frequently mutated in patients suffering from FAP and GS. APC has been found to be associated with structural components of intracellular junctions. b-catenin and g-catenin (also called plakoglobin) areMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol APC2
Antigen-navn Adenomatosis Polyposis Coli 2
Environmentally Preferable
Antigen synonymer anaphase-promoting complex subunit 2|APCL|cyclosome subunit 2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 10297
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR