Anti-APOBEC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-12492R-A555
BOSSBS-12492R-A555EA 10450 NOK
BOSSBS-12492R-A555
Anti-APOBEC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Post-transcriptional editing of apolipoprotein B (apoB) mRNA is regulated by This gene encodes a member of the cytidine deaminase enzyme family. The encoded protein forms a multiple-protein editing holoenzyme with APOBEC1 complementation factor (ACF) and APOBEC1 stimulating protein (ASP). This holoenzyme is involved in the editing of C-to-U nucleotide bases in apolipoprotein B and neurofibromatosis-1 mRNAs. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOBEC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol APOBEC1
Environmentally Preferable
Antigen-navn Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide 1
Antigen synonymer BEDP|CDAR1|APOBEC-1|HEPR
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 555
Reaktivitet Human
Vert Rabbit
Gen-ID 339
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR