Anti-APOBEC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-12492R-A647
BOSSBS-12492R-A647EA 10450 NOK
BOSSBS-12492R-A647
Anti-APOBEC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
Post-transcriptional editing of apolipoprotein B (apoB) mRNA is regulated by This gene encodes a member of the cytidine deaminase enzyme family. The encoded protein forms a multiple-protein editing holoenzyme with APOBEC1 complementation factor (ACF) and APOBEC1 stimulating protein (ASP). This holoenzyme is involved in the editing of C-to-U nucleotide bases in apolipoprotein B and neurofibromatosis-1 mRNAs. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOBEC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR