Anti-APOBEC2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverandør: Bioss
BS-12493R-A555
BOSSBS-12493R-A555EA 10450 NOK
BOSSBS-12493R-A555
Anti-APOBEC2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
APOBEC2 is a 224 amino acid protein that belongs to the cytidine and deoxycytidylate deaminase family. Expressed exclusively in heart and skeletal muscle, APOBEC2 is thought to be a probable C (cytidine) to U (uridine) editing enzyme. However, unlike other members of the family, such as APOBEC1, which mediates the editing of apolipoprotein (apo) B mRNA, APOBEC2 does not display any detectable apoB mRNA editing activity. Also, APOBEC2 has been shown to have low, but definite, intrinsic cytidine deaminase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOBEC2