Anti-APOBEC2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-12493R-A680
BOSSBS-12493R-A680EA 10425 NOK
BOSSBS-12493R-A680
Anti-APOBEC2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
APOBEC2 is a 224 amino acid protein that belongs to the cytidine and deoxycytidylate deaminase family. Expressed exclusively in heart and skeletal muscle, APOBEC2 is thought to be a probable C (cytidine) to U (uridine) editing enzyme. However, unlike other members of the family, such as APOBEC1, which mediates the editing of apolipoprotein (apo) B mRNA, APOBEC2 does not display any detectable apoB mRNA editing activity. Also, APOBEC2 has been shown to have low, but definite, intrinsic cytidine deaminase activity.

Type: Primary
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: ALEXAMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR