Anti-BLVRA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-12868R-A350
BOSSBS-12868R-A350EA 10450 NOK
BOSSBS-12868R-A350
Anti-BLVRA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
In human liver cytosolic fractions, four forms of biliverdin reductase have been identified, including two biliverdin-IX Beta reductases and two biliverdin-IX Alpha reductases, designated isozymes I and II and isozymes III and IV, respectively. Biliverdin reductase A (BLVRA), also designated biliverdin-IX Alpha-reductase, belongs to the GFO/iIDH/MocA family and the biliverdin reductase subfamily. The gene that encodes this cytoplasmic protein maps to chromosome 7p14-cen. BLVRA reduces biliverdin IX ?(the ?methene bridge of the open tetrapyrrole) to bilirubin with the concomitant oxidation ofMore Product Information