Anti-CRTC3 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-12950R-CY5
BOSSBS-12950R-CY5EA 10550 NOK
BOSSBS-12950R-CY5
Anti-CRTC3 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
TORC3 is a 619 amino acid protein that localizes to both the cytoplasm and the nucleus and belongs to the TORC family. Expressed in lung tissue and B and T lymphocytes, as well as in colon, brain, ovary, kidney, prostate, colon and heart, TORC3 functions as a transcriptional coactivator for CREB-1, thereby regulating the expression of CREB-activated genes, and is also thought to activate the SIK/TORC signaling pathway. TORC3 exists as multiple alternatively spliced isoforms and, in addition to its role in transcriptional activation, is thought to induce mitochondrial biogenesis, specificallyMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CRTC3
Environmentally Preferable
Antigen-navn CREB-Regulated Transcription Coactivator 3
Antigen synonymer TORC3|AI429792|BC011210|2610312F20Rik|6332415K15Rik
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 64784
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR