Anti-CRTC3 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-12950R-CY7
BOSSBS-12950R-CY7EA 10550 NOK
BOSSBS-12950R-CY7
Anti-CRTC3 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
TORC3 is a 619 amino acid protein that localizes to both the cytoplasm and the nucleus and belongs to the TORC family. Expressed in lung tissue and B and T lymphocytes, as well as in colon, brain, ovary, kidney, prostate, colon and heart, TORC3 functions as a transcriptional coactivator for CREB-1, thereby regulating the expression of CREB-activated genes, and is also thought to activate the SIK/TORC signaling pathway. TORC3 exists as multiple alternatively spliced isoforms and, in addition to its role in transcriptional activation, is thought to induce mitochondrial biogenesis, specificallyMore Product Information