Anti-DHX15 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-13001R
BOSSBS-13001REA 8500 NOK
BOSSBS-13001R
Anti-DHX15 Rabbit Polyclonal Antibody
Antistoffer
DEAD-box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure and ribosome/spliceosome assembly. Based on their distribution patterns, some members of this family may be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX15 (DEAH-box protein 15), also known as DHX15, DBP1 or HRH2, is a nuclear ATP-dependent RNA helicase that is involved in pre-mRNA splicing and is a member of the DEAH-box subfamily of DEAD-box proteins. ExpressedMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol DHX15
Environmentally Preferable
Antigen-navn DEAH (Asp-Glu-Ala-His) box helicase 15
Antigen synonymer PRP43|PRPF43|DBP1|PrPp43p|HRH2|DDX15
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 1665
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR