Anti-DNM1 Rabbit Polyclonal Antibody
Leverandør: Bioss
BS-13038R
BOSSBS-13038REA 8500 NOK
BOSSBS-13038R
Anti-DNM1 Rabbit Polyclonal Antibody
Antistoffer
Dynamin I is a GTPase enzyme required for the retrieval of synaptic vesicles after exocytosis and functions in endocytosis by stimulating assembly of invaginating synaptic vesicles (1). Dynamin I is phosphorylated in nerve terminals exclusively in the cytosolic compartment and in vitro by protein kinase C (PKC) (2–5). The phosphorylation site in PKC-phosphorylated Dynamin I is a single site at Serine 795, which is located near a binding site for the SH3 domain of p85, the regulatory subunit of phosphatidylinositol 3-kinase (2–5). Dephosphorylation is required for synaptic vesicleMore Product Information