Anti-EMSY Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-13069R-A647
BOSSBS-13069R-A647EA 10450 NOK
BOSSBS-13069R-A647
Anti-EMSY Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
EMSY binds BRCA2 within a region (exon 3) deleted in cancer. EMSY is capable of silencing the activation potential of BRCA2 exon 3, associates with chromatin regulators HP1 and BS69, and localizes to sites of repair following DNA damage. Its levels are amplified in breast and ovarian cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EMSY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR