Anti-GBAS Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-13299R-A647
BOSSBS-13299R-A647EA 10450 NOK
BOSSBS-13299R-A647
Anti-GBAS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
NIPSNAP2 is a 286 amino acid protein that is abundantly expressed in heart and skeletal muscle. Belonging to the NIPSNAP family, NIPSNAP2 may be involved in vesicular transport. NIPSNAP2 contains a signal peptide, a transmembrane domain and two tyrosine phosphorylation sites. NIPSNAP2 is encoded by a gene mapping to human chromosome 7p11.2. Chromosomal region 7p12 is amplified in approximately 40% of glioblastomas, the most common and malignant form of central nervous system tumor. Human chromosome 7 houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of theMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GBAS
Environmentally Preferable
Antigen-navn glioblastoma amplified sequence
Antigen synonymer NIPSNAP2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 647
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 2631
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR