Anti-MOGS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-13322R-A350
BOSSBS-13322R-A350EA 10450 NOK
BOSSBS-13322R-A350
Anti-MOGS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Glycosylation of asparagine residues in Asn-X-Ser/Thr motifs in proteins commonly occur in the lumen of the endoplasmic reticulum (ER). Glucosidase I catalyzes the first step in the N-linked oligosaccharide processing pathway. It specifically removes the distal alpha 1,2-linked glucose residue from the Glc3-Man9-GlcNAc2 oligosaccharide precursor. Glucosidase I contains a short cytosolic tail, a single pass transmembrane domain and a large C-terminal catalytic domain located on the luminal side of the ER. Mutations in the gene encoding Glucosidase I result in the congenital disorderMore Product Information