Anti-HESX1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-13597R-A488
BOSSBS-13597R-A488EA 10450 NOK
BOSSBS-13597R-A488
Anti-HESX1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
The homeobox protein, HESX1, which is also known as Rathke’s pouch homeobox, HANF, homeodomain transcription factor, and anterior-restricted homeobox protein is a transcription factor that belongs to the homeodomain family of DNA binding proteins. HESX1 is initially expressed in embryonic stem cells and the primitive forebrain, and is essential for normal development of the eyes and other anterior CNS structures, such as the hypothalamus, the pituitary gland and the olfactory bulbs. The homeobox gene Hesx1 is expressed in the anterior visceral endoderm (AVE), anterior axial mesendoderm (More Product Information