Anti-HESX1 Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-13597R-CY5
BOSSBS-13597R-CY5EA 10550 NOK
BOSSBS-13597R-CY5
Anti-HESX1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
The homeobox protein, HESX1, which is also known as Rathke’s pouch homeobox, HANF, homeodomain transcription factor, and anterior-restricted homeobox protein is a transcription factor that belongs to the homeodomain family of DNA binding proteins. HESX1 is initially expressed in embryonic stem cells and the primitive forebrain, and is essential for normal development of the eyes and other anterior CNS structures, such as the hypothalamus, the pituitary gland and the olfactory bulbs. The homeobox gene Hesx1 is expressed in the anterior visceral endoderm (AVE), anterior axial mesendoderm (More Product Information