Anti-AIF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-1363R-A680
BOSSBS-1363R-A680EA 10525 NOK
BOSSBS-1363R-A680
Anti-AIF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Actin-binding protein that enhances membrane ruffling and RAC activation. Enhances the actin-bundling activity of LCP1. Binds calcium. Plays a role in RAC signaling and in phagocytosis. May play a role in macrophage activation and function. Promotes the proliferation of vascular smooth muscle cells and of T-lymphocytes. Enhances lymphocyte migration. Plays a role in vascular inflammation.

Type: Primary
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-147/147
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR