Anti-AIF1 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-1363R-CY5
BOSSBS-1363R-CY5EA 10650 NOK
BOSSBS-1363R-CY5
Anti-AIF1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Actin-binding protein that enhances membrane ruffling and RAC activation. Enhances the actin-bundling activity of LCP1. Binds calcium. Plays a role in RAC signaling and in phagocytosis. May play a role in macrophage activation and function. Promotes the proliferation of vascular smooth muscle cells and of T-lymphocytes. Enhances lymphocyte migration. Plays a role in vascular inflammation.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(IHC-F)(1:50-200

Type: Primary
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR