Anti-GRB7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-13642R-FITC
BOSSBS-13642R-FITCEA 10550 NOK
BOSSBS-13642R-FITC
Anti-GRB7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Many growth factors function by binding receptors with intrinsic tyrosine kinase activity (1,2). Signaling by such receptors involves a series of intermediates characterized by SH2 domains that bind tyrosine phosphorylated receptors by a direct interaction between the SH2 domain and the phosphotyrosine-containing receptor sequences (3,4). GRB7, a SH2 domain protein, has a single SH2 domain at its C-terminal, a central region with similarity to Ras GAP and a proline-rich N-terminus (5,6). GRB7 maps to the region on mouse chromosome 11 containing the Neu gene (6). This region of mouseMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GRB7
Antigen-navn Growth Factor Receptor-bound Protein 7
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR