Anti-Claudin 23 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-13746R-A350
BOSSBS-13746R-A350EA 10450 NOK
BOSSBS-13746R-A350
Anti-Claudin 23 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
The claudin superfamily consists of structurally related proteins that are important structural and functional components of tight junctions. Claudin-23, also known as CLDN23 or CLDNL, is a 292 amino acid multi-pass membrane protein that localizes to cell junctions and belongs to the claudin family. Expressed in stomach and placenta, as well as in germinal center B-cells, claudin-23 is thought to exhibit calcium-dependent cell-adhesion activity through which it plays an essential role in tight junction-specific obliteration of the intercellular space. Human claudin-23 shares 80% sequenceMore Product Information