Anti-Claudin 23 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-13746R-A750
BOSSBS-13746R-A750EA 10425 NOK
BOSSBS-13746R-A750
Anti-Claudin 23 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
The claudin superfamily consists of structurally related proteins that are important structural and functional components of tight junctions. Claudin-23, also known as CLDN23 or CLDNL, is a 292 amino acid multi-pass membrane protein that localizes to cell junctions and belongs to the claudin family. Expressed in stomach and placenta, as well as in germinal center B-cells, claudin-23 is thought to exhibit calcium-dependent cell-adhesion activity through which it plays an essential role in tight junction-specific obliteration of the intercellular space. Human claudin-23 shares 80% sequenceMore Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Claudin 23
Antigen synonymer Claudin23|2310014B08Rik|CLDNL|CLDN 23|CLDN23
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Human
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 25-81/292
Immunogen KLH conjugated synthetic peptide derived from human Claudin 23
Skjema Liquid
Molekylvekt 32 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR