Anti-Claudin 16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-13752R-A750
BOSSBS-13752R-A750EA 10425 NOK
BOSSBS-13752R-A750
Anti-Claudin 16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Plays a major role in tight junction-specific oblitreation of the intercellular space, through calcium-independent cell-adhesion activity. Involved in paracellular magnesium reabsorption. Required for a selective paracellular conductance. May form, alone or in partnership with other constituents, an intercellular pore permitting paracellular passage of magnesium and calcium ions down their electrochemical gradients. Alternatively, it could be a sensor of magnesium concentration that could alter paracellular permeability mediated by other factors.

Type: Primary
Antigen: Claudin 16
Clonality:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR