Anti-Arfaptin 2 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-1442R-CY5
BOSSBS-1442R-CY5EA 10550 NOK
BOSSBS-1442R-CY5
Anti-Arfaptin 2 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
ARFIP2 is a ubiquitously expressed protein implicated in mediating cross talk between RAC and ARF small GTPases. It has been shown that ARFIP2 binds specifically to GTP-bound ARF1 and ARF6, but binds to Rac-GTP and Rac-GDP with similar affinities. The X-ray structure of arfaptin reveals an elongated, crescent-shaped dimer of 3-helix coiled-coils. Structures of arfaptin with Rac bound to either GDP or the slowly hydrolysable analog GMPPNP show that the switch regions adopt similar conformations in both complexes.

Type: Primary
Antigen: Arfaptin 2
Clonality: Polyclonal
Clone:
Conjugation: Cy5
More Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn Arfaptin 2
Antigen synonymer ARFP2_HUMAN .|arfaptin 2|POR1|ADP-ribosylation factor interacting protein 2|Partner of RAC1|ARFIP2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Vert Rabbit
Gen-ID 23647
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 221-341/341
Immunogen KLH conjugated synthetic peptide derived from human Arfaptin 2
Skjema Liquid
Molekylvekt 38 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR