Anti-Promotilin Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-1445R
BOSSBS-1445REA 8500 NOK
BOSSBS-1445R
Anti-Promotilin Rabbit Polyclonal Antibody
Antistoffer
Plays an important role in the regulation of interdigestive gastrointestinal motility and indirectly causes rhythmic contraction of duodenal and colonic smooth muscle.

Type: Primary
Antigen: Promotilin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Public Immunogen Range: 51-115/115
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR