Anti-RGS14 Rabbit Polyclonal Antibody
Leverandør: Bioss
BS-15502R
BOSSBS-15502REA 8500 NOK
BOSSBS-15502R
Anti-RGS14 Rabbit Polyclonal Antibody
Antistoffer
RGS14 is a member of the regulator of G-protein signaling family. This protein contains one RGS domain, two Raf-like Ras-binding domains (RBDs), and one GoLoco domain. The protein attenuates the signaling activity of G-proteins by binding, through its GoLoco domain, to specific types of activated, GTP-bound G alpha subunits. Acting as a GTPase activating protein (GAP), the protein increases the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.More Product Information