Anti-IDH3B Rabbit Polyclonal Antibody
Leverandør: Bioss
BS-15538R
BOSSBS-15538REA 8500 NOK
BOSSBS-15538R
Anti-IDH3B Rabbit Polyclonal Antibody
Antistoffer
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of theMore Product Information