Anti-IDH3B Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Leverandør: Bioss
BS-15538R-A680
BOSSBS-15538R-A680EA 10425 NOK
BOSSBS-15538R-A680
Anti-IDH3B Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antistoffer
Isocitrate dehydrogenases catalyse the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilises NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localise to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyse the allosterically regulated rate-limiting step of theMore Product Information