Anti-ENPP3/CD203c Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: Bioss
BS-1568R-PE
BOSSBS-1568R-PEEA 10775 NOK
BOSSBS-1568R-PE
Anti-ENPP3/CD203c Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Cleaves a variety of phosphodiester and phosphosulfate bonds including deoxynucleotides, nucleotide sugars, and NAD.

Type: Primary
Antigen: ENPP3/CD203c
Clonality: Polyclonal
Clone:
Conjugation: PE
Public Immunogen Range: 40-140/875
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR