Anti-APOH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-1570R-FITC
BOSSBS-1570R-FITCEA 10550 NOK
BOSSBS-1570R-FITC
Anti-APOH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Binds to various kinds of negatively charged substances such as heparin, phospholipids, and dextran sulfate. May prevent activation of the intrinsic blood coagulation cascade by binding to phospholipids on the surface of damaged cells.

Type: Primary
Antigen: APOH
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 281-345/345
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn APOH
Antigen synonymer B2GPI|Activated protein C-binding protein|APC inhibitor|Anticardiolipin cofactor|BG|APOH|B2G1|B2GP1|Apolipoprotein H|Beta-2-glycoprotein I|Beta(2)GPI|Apo-H|Beta-2-glycoprotein 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 350
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 281-345/345
Immunogen KLH conjugated synthetic peptide derived from human ApoH
Skjema Liquid
Molekylvekt 36 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR