Anti-IFNA8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-1578R-A488
BOSSBS-1578R-A488EA 10450 NOK
BOSSBS-1578R-A488
Anti-IFNA8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.

Type: Primary
Antigen: IFNA8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 161-189/189
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn IFNA8
Antigen synonymer LeIF B|IFN-alphaB|Interferon alpha-8|Interferon alpha-B2|IFN-alpha-8|Interferon alpha-B
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 488
Reaktivitet Human
Vert Rabbit
Gen-ID 3445
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Modifikasjon Unmodified
Epitop 161-189/189
Immunogen KLH conjugated synthetic peptide derived from human IFNA8
Skjema Liquid
Molekylvekt 21 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR