Anti-Group I mGLUR Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-1803R-CY7
BOSSBS-1803R-CY7EA 10650 NOK
BOSSBS-1803R-CY7
Anti-Group I mGLUR Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activateMore Product Information